missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COPS8 Recombinant Rabbit Monoclonal Antibody (23GB3100), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | COPS8 |
|---|---|
| Clone | 23GB3100 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088433
![]() |
Invitrogen™
MA552544 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088110
|
Invitrogen™
MA552545 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COPS8 Recombinant Monoclonal antibody specifically detects COPS8 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| COPS8 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q6P4Z9, Q8VBV7, Q99627 | |
| 108679, 10920, 363283 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3100 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 9430009J09Rik; AA408242; COP9; COP9 (constitutive photomorphogenic) homolog, subunit 8; COP9 (constitutive photomorphogenic) homolog, subunit 8 (Arabidopsis thaliana); COP9 constitutive photomorphogenic homolog subunit 8; COP9 homolog; COP9 signalosome complex subunit 8; COP9 signalosome subunit 8; COPS8; Csn8; hCOP9; JAB1-containing signalosome subunit 8; SGN8; signalosome subunit 8 | |
| A synthesized peptide derived from CSN8 (180-209AA). | |
| Primary | |
| -20°C | |
| Cops8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title