missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | COPS8 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227872
|
Novus Biologicals
NBP3-35268-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229058
|
Novus Biologicals
NBP3-35268-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COPS8 Polyclonal antibody specifically detects COPS8 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| COPS8 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 10920 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| COP9, COP9 constitutive photomorphogenic homolog subunit 8 (Arabidopsis), COP9 homolog, COP9 signalosome complex subunit 8, CSN8JAB1-containing signalosome subunit 8, hCOP9, MGC1297, MMMEP, SGN8MGC43256, Signalosome subunit 8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-209 of human COPS8 (NP_006701.1).,, Sequence:, MPVAVMAESAFSFKKLLDQCENQELEAPGGIATPPVYGQLLALYLLHNDMNNARYLWKRIPPAIKSANSELGGIWSVGQRIWQRDFPGIYTTINAHQWSETVQPIMEALRDATRRRAFALVSQAYTSIIADDFAAFVGLPVEEAVKGILEQGWQADSTTRMVLPRKPVAGALDVSFNKFIPLSEPAPVPPIPNEQQLARLTDYVAFLEN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title