missing translation for 'onlineSavingsMsg'
Learn More

COPS8 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18411416 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411416 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411416 Supplier Novus Biologicals Supplier No. NBP176373JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COPS8 Polyclonal antibody specifically detects COPS8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPS8
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias COP9, COP9 constitutive photomorphogenic homolog subunit 8 (Arabidopsis), COP9 homolog, COP9 signalosome complex subunit 8, CSN8JAB1-containing signalosome subunit 8, hCOP9, MGC1297, MMMEP, SGN8MGC43256, Signalosome subunit 8
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human CSN8. The immunogen is located within the last 50 amino acids of CSN8. Amino Acid Squence: PLSEPAPVPPIPNEQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10920
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.