missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COPS6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544972
This item is not returnable.
View return policy
Description
Peptide sequence: DHVARMTATG SGENSTVAEH LIAQHSAIKM LHSRVKLILE YVKASEAGEV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
The protein encoded by this gene is one of the eight subunits of COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases. This protein belongs to translation initiation factor 3 superfamily. It is involved in the regulation of cell cycle and likely to be a cellular cofactor for HIV-1 accessory gene product Vpr.
Specifications
| COPS6 | |
| Polyclonal | |
| Unconjugated | |
| COPS6 | |
| COP9 (constitutive photomorphogenic) homolog, subunit 6; COP9 (constitutive photomorphogenic) homolog, subunit 6 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 6; COP9 complex S6; COP9 constitutive photomorphogenic homolog subunit 6; COP9 signalosome complex subunit 6; COP9 signalosome complex subunit 6-like protein; COP9 signalosome subunit 6; cops6; CSN6; H_NH0506M12.12; hVIP; hypothetical protein LOC550465; JAB1-containing signalosome subunit 6; MOV34 homolog; MOV34 homolog, 34 kD; MOV34-34KD; Sgn3; SGN6; signalosome subunit 6; VIP/MOV34; Vpr-interacting protein; zgc:112075 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10980 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7L5N1 | |
| COPS6 | |
| Synthetic peptide directed towards the middle region of human COPS6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction