missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS6, Mouse anti-Human, Clone: 2H5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010980-M08.100ug
This item is not returnable.
View return policy
Description
Sequence: VMACGVTGSVSVALHPLVILNISDHWIRMRSQEGRPVQVIGALIGKQEGRNIEVMNSFELLSHTVEEKIIIDKEYYYTKEEQFKQVFKELEFLGWYTTGGPPDPSDIHVHKQVCEIIESPLFLKLNPMTKHTDLPVSVFESVIDIINGEATMLFAELTYTLATEEAERIGVDHVARMTATGSGENSTVAEHLIAQHSAIKMLHSRVKLILEYVKASEAGEVPFNHEILREAYALCHCLPVLSTDKFKTDFYDQCNSpecifications
| COPS6 | |
| Monoclonal | |
| Unconjugated | |
| COP9 constitutive photomorphogenic homolog subunit 6 (Arabidopsis) | |
| CSN6/MOV34-34KD | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin) | |
| 2H5 | |
| In 1x PBS, pH 7.4 | |
| BC002520 | |
| COPS6 | |
| COPS6 (AAH02520, 30 a.a. ∼ 327 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10980 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction