missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COPS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557863
This item is not returnable.
View return policy
Description
Immunogen sequence: ALKHALHCTI LASAGQQRSR MLATLFKDER CQQLAAYGIL EKMYLDRIIR GNQLQEFAAM LMPHQKATTA DGSSILDRAV IEHNLLSASK LY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes one of eight subunits composing COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| COPS4 | |
| Polyclonal | |
| Unconjugated | |
| COPS4 | |
| AW208976; COP9 (constitutive photomorphogenic) homolog, subunit 4; COP9 (constitutive photomorphogenic) homolog, subunit 4 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 4; COP9 complex S4; COP9 constitutive photomorphogenic homolog subunit 4; COP9 constitutive photomorphogenic-like protein subunit 4; COP9 signalosome complex subunit 4; COP9 signalosome subunit 4; COPS4; CSN4; D5Ertd774e; JAB1-containing signalosome subunit 4; SGN4; signalosome subunit 4; testis tissue sperm-binding protein Li 42a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 360915, 51138 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68FS2, Q9BT78 | |
| COPS4 | |
| Recombinant protein corresponding to Human COPS4. Recombinant protein control fragment (Product #RP-96659). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction