missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS4 Antibody (SR1873), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | COPS4 |
|---|---|
| Clone | SR1873 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622025
|
Novus Biologicals
NBP3-22524-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611955
|
Novus Biologicals
NBP3-22524-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COPS4 Monoclonal antibody specifically detects COPS4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| COPS4 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| COP9 constitutive photomorphogenic homolog subunit 4 (Arabidopsis), COP9 signalosome complex subunit 4, CSN4Arabidopsis, homolog) subunit 4, MANOP1, MGC10899, MGC15160 | |
| A synthesized peptide derived from human COPS4 (Uniprot #: Q9BT78) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1873 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Neuronal Cell Markers, Neurotransmission | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 51138 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title