missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS2, Mouse anti-Human, Clone: 4B12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009318-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: REHIEELLRNIRTQVLIKLIKPYTRIHIPFISKELNIDVADVESLLVQCILDNTIHGRIDQVNQLLELDHQKRGGARYTALDKWTNQLNSLNQAVVSKLASpecifications
| COPS2 | |
| Monoclonal | |
| Unconjugated | |
| COP9 constitutive photomorphogenic homolog subunit 2 (Arabidopsis) | |
| ALIEN/CSN2/SGN2/TRIP15 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4B12 | |
| In 1x PBS, pH 7.4 | |
| NM_004236 | |
| COPS2 | |
| COPS2 (NP_004227, 344 a.a. ∼ 443 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9318 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction