missing translation for 'onlineSavingsMsg'
Learn More

Coproporphyrin III, MedChemExpress

Product Code. 16420759
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16420759 1 mg 1mg
16416077 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16420759 Supplier Medchem Express Supplier No. HY101398
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Coproporphyrin III (Zincphyrin) is a naturally occurring porphyrin derivative that is mainly found in urine.

Coproporphyrin III (Zincphyrin) is a naturally occurring porphyrin derivative that is mainly found in urine.

Specifications

Chemical Name or Material Coproporphyrin III
CAS 14643-66-4
Color Brown
Purity Grade Notes Research
Molecular Formula C36H38N4O8
Quantity 1 mg
Synonym Zincphyrin
Solubility Information DMSO : ≥ 83.3 mg/mL (127.23 mM)
SMILES O=C(O)CCC1=C2/C=C3C(CCC(O)=O)=C(C)C(/C=C(N/4)/C(C)=C(CCC(O)=O)C4=C\C5=N/C(C(CCC(O)=O)=C5C)=C\C(N2)=C1C)=N/3
Molecular Weight (g/mol) 654.71
Formula Weight 654.71
Percent Purity 96.92%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.