missing translation for 'onlineSavingsMsg'
Learn More

Coproporphyrin I-15N4, TRC

Product Code. 30012903
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012903 5 mg 5mg
30004328 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012903 Supplier Toronto Research Chemicals Supplier No. TRCC6854015MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C36H3815N4O8
Molecular Weight (g/mol) 658.68
Synonym 3,8,13,18-Tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropanoic -15N4 Acid, 3,8,13,18-Tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropionic-15N4 Acid, NSC 267073-15N4
IUPAC Name 3-[7,12,17-tris(2-carboxyethyl)-3,8,13,18-tetramethyl-21,22-dihydroporphyrin-2-yl]propanoic acid
SMILES CC(C(/C=C(C(CCC(O)=O)=C/1C)\[15NH]C1=C/2)=[15N]/3)=C(CCC(O)=O)C3=C\C4=C(C)C(CCC(O)=O)=C(/C=C5C(C)=C(CCC(O)=O)C2=[15N]/5)[15NH]4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C36H38N4O8/c1-17-21(5-9-33(41)42)29-14-26-19(3)23(7-11-35(45)46)31(39-26)16-28-20(4)24(8-12-36(47)48)32(40-28)15-27-18(2)22(6-10-34(43)44)30(38-27)13-25(17)37-29/h13-16,37-38H,5-12H2,1-4H3,(H,41,42)(H,43,44)(H,45,46)(H,47,48)/i37+1,38+1,39+1,40+1
Percent Purity >95%
Chemical Name or Material Coproporphyrin I-15N4
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.