missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coprelotamab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Coprelotamab Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284128
GREENER_CHOICE
View Documents
Invitrogen™
MA559195
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282553
View Documents
Invitrogen™
MA559196
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Coprelotamab is a biosimilar monoclonal antibody directed against the human epidermal growth factor receptor 2 (HER2), also known as ERBB2. The ERBB2 gene, located on chromosome 17 at position 17q12, encodes the ErbB2 protein, also known as HER2 (human epidermal growth factor receptor 2). The ErbB2 protein is a member of the receptor tyrosine kinase family and is characterized by its extracellular domain, transmembrane segment, and intracellular tyrosine kinase domain. Structurally, the extracellular domain of ErbB2 forms a homodimer through a 'back to head' interaction, which is critical for its function. ERBB2 is best known for its role in the pathogenesis of various cancers, particularly breast cancer. Overexpression or amplification of the ERBB2 gene leads to excessive ErbB2 protein on the cell surface, resulting in constant cell growth signaling. Unlike other members of the ErbB family, no specific ligand has been found for ErbB2, and its activation often involves heterodimerization with other ErbB family receptors such as EGFR. The activation of these receptors triggers downstream signaling pathways that promote cell proliferation and survival.
Specifications

Specifications

Coprelotamab Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.