missing translation for 'onlineSavingsMsg'
Learn More

Copper wire, 0.025mm (0.001in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 11371548
Click to view available options
Quantity:
10 m
200 m
50 m
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 m
Percent Purity 99.995%
Grade Puratronic™
Odor Odorless
Weight ≈0.0044g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Copper wire, 0.025mm (0.001 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'