missing translation for 'onlineSavingsMsg'
Learn More

Copper tubing, 6.35mm (0.25in) OD, 4.72mm (0.186in) ID, Thermo Scientific™

Product Code. 15417075
Click to view available options
Quantity:
3 m
Unit Size:
3m
This item is not returnable. View return policy

Product Code. 15417075

Brand: Thermo Scientific Alfa Aesar 045793.G4

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7440-50-8
Quantity 3 m
Odor Odorless
Molecular Formula Cu
MDL Number MFCD00010965
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
IUPAC Name copper
Molecular Weight (g/mol) 63.55
PubChem CID 23978
ChEBI CHEBI:30052
Chemical Name or Material Copper tubing, OD: 6.35 mm (0.25 in.), ID: 4.72 mm (0.186 in.)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.