missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Tin Bronze Navy G, CDA 903, C90300, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Tin Bronze Navy G, CDA 903, C90300, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Silicon/Arsenic/Iron/Phosphorus/Silver/Oxygen/Antimony/Nickel/Lead/Zinc/Tin/Copper/Nitrogen/Tellurium/Beryllium/Cadmium/Cobalt/Manganese/Selenium/Zirconium/Aluminium/Chromium/Niobium/Bismuth |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon 0.002 %/Silicon 0.003 %/Arsenic 0.004 %/Iron 0.004 %/Phosphorus 0.004 %/Silver 0.005 %/Oxygen 0.006 %/Antimony 0.008 %/Nickel 0.008 %/Lead 0.17 %/Zinc 3 %/Tin 9.14 %/Copper 87.5 %/Nitrogen (0.0004) %/Tellurium (0.0004) %/Beryllium (0.0005) %/Cadmium (0.001) %/Cobalt (0.001) %/Manganese (0.001) %/Selenium (0.001) %/Zirconium (0.001) %/Aluminium (0.002) %/Chromium (0.002) %/Niobium (0.005) %/Bismuth (0.01) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction