missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Copper Test Kits

Product Code. 11771073
Click to view available options
No. of Tests:
19
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11771073

Brand: WTW™ 250408

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Analytes Cuprizon
For Use With (Application) For waste water, drinking water, ground water, foodstuffs
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex, Photoflex photometers
Includes Certificates
No. of Tests 19
Sufficient For 25 Tests
Test Method Photometric
Type Copper (Cu) cell test kit
Hazard Statement
  • EUH210-Safety data sheet available on request.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.