missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, 0.0200 M (M/50), Ricca Chemical

Product Code. 31750111
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750111 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750111 Supplier Ricca Chemical Company Supplier No. R23492001A
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aqueous solution designed for use as a general laboratory reagent.

Specifications

CAS 7758-99-8, 7732-18-5
Color Blue
CAS Min % 98.0, 0.49
pH ∼4
CAS Max % 100.0, 0.51
Physical Form Liquid
Quantity 1 L
Packaging Natural Poly Bottle
Molecular Formula CuH10O9S
MDL Number MFCD00149681
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
IUPAC Name copper(2+) pentahydrate sulfate
Molecular Weight (g/mol) 249.68
PubChem CID 24463
ChEBI CHEBI:31440
Grade Laboratory
Chemical Name or Material Copper Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.