missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper for AAS 1g/L HNO3 matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£23.60 - £63.65
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | HNO3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12973265
|
Fisher Chemical
B 115 100 |
100 mL |
£23.60
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12993265
|
Fisher Chemical
B 115 500 |
500 mL |
£43.20
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12983265
|
Fisher Chemical
B1151000 |
1000 mL | |||||||
Specifications
1g/L | |
HNO3 |
Safety and Handling
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for AAS. Copper (Cu) 1000mg/l in HNO3 2%
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .