missing translation for 'onlineSavingsMsg'
Learn More

Copper Standard Metal Solution 1000ppm, for ICP, Solution HNO3 1M, Fisher Chemical

Product Code. 10724951 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10724951 500 mL HDPE plastic bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10724951 Supplier Fisher Chemical Supplier No. J/8256/08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3251-23-8 | CuN2O6 | 187.554 g/mol

500mL, HDPE plastic bottle

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
pH 1
CAS 3251-23-8
Quantity 500 mL
Packaging HDPE plastic bottle
Form Liquid
Molecular Formula CuN2O6
MDL Number 10967
Synonym copper dinitrate, cupric nitrate, copper ii nitrate, copper nitrate, cupric dinitrate, claycop, copper 2+ nitrate, caswell no. 246, nitric acid, copper 2+ salt, unii-9tc879s2zv
InChI Key XTVVROIMIGLXTD-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
IUPAC Name copper;dinitrate
Molecular Weight (g/mol) 187.554
PubChem CID 18616
ChEBI CHEBI:78036
Show More Show Less
compliance-icons
Product Identifier
  • Copper solution 1000 ppm, standard metal solution in approx. 1M nitric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Nitric acid, Cupricnitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.