missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Standard for ICP, ICP-MS 10,000 μg/mL (10,000 pm) in 2-5% Nitric Acid (HNO3), Reagecon
Shop All Reagecon Products

Description
- ICP, ICP-MS Reference Standard, ICP, ICP-MS Reference Materia
- ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
- ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
- ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
- ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
- ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
- GHS/OSHA compliant, REACH etc.
- This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), triple distilled acids and triple leached pre-cleaned bottles.
- The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
- All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.
Specifications
Specifications
| Description | Matrix: 2-5% HNO3 |
| Packaging | HDPE Triple-leached |
| Analyte or Component Names | Copper |
| Physical Form | Liquid |
| Concentration | 10,000 µg/mL (10,000 ppm) |
| Technique | ICP, ICP-MS, AA, Ion Chromatography, XRF, Cation Measurement |
| Element | Cu |
| Quantity | 250 mL |
Safety and Handling
Product Identifier
- COPPER ICP STANDARD SOLUTION
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contain: nitric acid, copper
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?