missing translation for 'onlineSavingsMsg'
Få mere at vide

Copper Standard: Cu @ 10000 μg/mL in 5% HNO3

Artikelnummer. 30312973
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
100 mL
500 mL
Pakningsstørrelse:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
30312973 500 mL 500mL
30327219 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Artikelnummer. 30312973 Leverandør VHG Labs Leverandørnr. VHGTCUN500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper
CAS 7440-50-8
Analyte or Component Names Copper
Shipping Condition Room Temperature
Molecular Formula Cu
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Copper, Cuprum, Blister copper, Cathode copper, Copper powder
InChI Formula InChI=1 S/Cu
Solution Type Single Solution
SMILES [Cu]
Element Cu
IUPAC Name copper
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 63.55
Quantity 500 mL
Formula Weight 62.9296
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.