missing translation for 'onlineSavingsMsg'
Learn More

Copper solution for ICP 10g/L HNO3 matrix, Fisher chemical™
BIOPHARMA

Product Code. 12976284 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12976284 500 mL Bottle 500mL
12956284 100 mL Bottle 100mL
12966284 250 mL Bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12976284 Supplier Fisher Chemical Supplier No. S 215 500
  • ONLINE EXCLUSIVE PRICE
    £270.77
    £216.00 / 500mL
    Save £54.77
    20% Off
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Tekniske data

Packaging Bottle
Analyte or Component Names Cu
Concentration 10±0.02 g/L
Type Mass Spectrometry Standard
Element Cu
Solvent or Matrix Nitric Acid
Quantity 500 mL
compliance-icons compliance-icons
Produktidentifikator
  • Single-element Standard Solution for ICP. Copper (Cu) 10000mg/l in HNO3 5%
Signalord
  • Danger
Farekategori
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Faresætning
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Sikkerhedssætning
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.