missing translation for 'onlineSavingsMsg'
Learn More

Copper silicide, 99.5% (metals basis), Thermo Scientific Chemicals

£82.00 - £214.00

Chemical Identifiers

CAS 12159-07-8
Molecular Formula Cu5Si
MDL Number MFCD00134719
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357577
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012844.14
25 g
£82.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11367577
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012844.22
100 g
£214.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Copper silicide thin film is used for passivation of copper-based chips, where it serves to suppress diffusion and electromigration and serves as a diffusion barrier. It is involved in the direct process, the industrial route to organosilicon compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12159-07-8
Cu5Si
MFCD00134719

Specifications

825°C
Silver
99.5% (Metals basis)
MFCD00134719
345.79
Copper silicide
12159-07-8
Pieces
Cu5Si
Insoluble in water.
Odorless
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 235-286-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only