missing translation for 'onlineSavingsMsg'
Learn More

Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11331218
Click to view available options
Quantity:
500 g
2 kg
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 2 kg
Percent Purity 99.9%
Odor Odorless
Form Shot
Assay (metals basis)
Chemical Name or Material Copper shot, 1 to 10mm (0.04 to 0.4 in.)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'