missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper powder, spherical, -100+325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Appearance | Copper colored powder |
Assay Percent Range | 99.9% (Metals basis) |
Quantity | 2 kg |
Mesh Size | -100+325 Mesh |
Odor | Odorless |
Vapor Pressure | 0 hPa |
Form | Spherical Powder |
Chemical Name or Material | Copper |
Safety and Handling
Product Identifier
- Copper powder
- Warning
- Flammable solid Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'