missing translation for 'onlineSavingsMsg'
Learn More

Copper plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 6.35mm (0.25in) thick

Product Code. 15453955
Change view
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453955 150 x 150 mm Each
15463955 300 x 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453955 Supplier Thermo Scientific Alfa Aesar Supplier No. 045190.GT
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7440-50-8 | Cu | 63.55 g/mol

Copper foil, Oxygen-Free High Conductivity (OFHC), alloy 101 is used for automotive rectifiers, transistor component bases, high resistance-ratio cryogenic shunts, wave guides, hollow conductors, anodes for vacuum tubes, coaxial cable, coaxial tube, klystrons, microwave tubes, bus bars, lead-in wire, vacuum seals and glass-to-metal seals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
SMILES [Cu]

Specifications

Appearance Copper colored plate
Quantity 150 x 150 mm
Odor Odorless
Form Plate
Solubility Information Insoluble in water.
IUPAC Name nitric acid
Chemical Name or Material Copper

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.