missing translation for 'onlineSavingsMsg'
Learn More

Copper, plasma standard solution, Specpure™ Cu 10,000μg/mL, Thermo Scientific Chemicals

£63.00 - £328.00

Chemical Identifiers

MDL Number MFCD00010965
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307937
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014372.AD
50 mL
£63.00
50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11397927
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014372.AE
100 mL
£101.00
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11440410
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014372.AP
500 mL
£328.00
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00010965

Specifications

Cu in 5% HNO3
UN3264
Liquid
MFCD00010965
Specpure™
Copper plasma standard solution, Cu 10000μg/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-714-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only