missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Phosphor Bronze, ASTM B888, C52480, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Phosphor Bronze, ASTM B888, C52480, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Bismuth/Silicon/Phosphorus/Silver/Antimony/Iron/Lead/Nickel/Zinc/Tin/Copper |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Bismuth (0.0015) %/Silicon (0.002) %/Phosphorus (0.011) %/Silver (0.015) %/Antimony (0.018) %/Iron (0.02) %/Lead (0.325) %/Nickel (0.36) %/Zinc (0.39) %/Tin (10.4) %/Copper (88.5) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction