missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Phosphor Bronze, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Phosphor Bronze, 50g
Specifications
Specifications
| Chemical Name or Material | Aluminium/Iron/Arsenic/Silicon/Zinc/Antimony/Silver/Bismuth/Lead/Nickel/Tin/Copper |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Aluminium (0.001) %/Iron (0.002) %/Arsenic (0.003) %/Silicon (0.005) %/Zinc (0.008) %/Antimony (0.016) %/Silver (0.017) %/Bismuth (0.053) %/Lead (0.087) %/Nickel (0.125) %/Tin (17.6) %/Copper (82) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction