missing translation for 'onlineSavingsMsg'
Learn More

Copper Nickel wire, 0.5mm (0.02in) dia

Product Code. 11389378
Change view
Click to view available options
Quantity:
100 m
25 m
500 m
Unit Size:
100m
25m
500m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389378 100 m 100m
11399378 25 m 25m
11309388 500 m 500m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389378 Supplier Thermo Scientific Alfa Aesar Supplier No. 043272.H4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Copper Nickel wire is employed for sea water condenser systems and in desalination plants, as well as pipe work in chemical plants. It is being increasingly employed for brake and hydraulic suspension systems and cooling systems in cars and commercial vehicles. It is commonly used for the manufacture of coins, medals and other semi valuable applications. It is also used in cooling circuits, ammunition, sea water corrosion-resistant assemblies and condenser tubes. The good resistance to biofouling and sea water corrosion resistance of the copper nickel alloys have lead to its use in applications such as cladding for ships' hulls, legs of oil rig platforms and sea water intake screens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Nickel wire
Name Note 0.5mm (0.02 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Ni 55:45 wt%
Quantity 100 m
Weight ≈1.75g/m
Form Wire
MDL Number MFCD00801098
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.