Learn More
Copper Nickel wire, 0.075mm (0.003in) dia, Thermo Scientific Chemicals
Resistance alloy, negative lead for type J thermocouple
Brand: Thermo Scientific Alfa Aesar 043269.H4
16.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Resistance alloy, negative lead for type J thermocouple
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper Nickel wire | |
Cu:Ni 55:45 wt% | |
100 m | |
≈0.04g/m |
0.075mm (0.003 in.) dia. | |
MFCD00801098 | |
Constantan | |
Wire |
RUO – Research Use Only