missing translation for 'onlineSavingsMsg'
Learn More

Copper Manganese Nickel wire, 0.1mm (0.004 in.) dia., Thermo Scientific™

Product Code. p-7058758
Click to view available options
Quantity:
100 m
500 m
Unit Size:
100m
500m
This item is not returnable. View return policy

Product Code. 11334888

Brand: Thermo Scientific Alfa Aesar 043188.H4

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Resistance alloy for thermocouples and electronics industry

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Manganese Nickel wire
Name Note 0.1mm (0.004 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Mn:Ni; 84:12:4 wt%
Quantity 100 m
Weight ≈0.066g/m
Form Wire
MDL Number MFCD01866121
Synonym Manganin™
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.