missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Leaded Red Brass, CDA 836, C83600, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Leaded Red Brass, CDA 836, C83600, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Cadmium/Selenium/Niobium/Cobalt/Arsenic/Phosphorus/Silver/Bismuth/Sulfur/Iron/Antimony/Nickel/Lead/Tin/Zinc/Copper/Oxygen/Manganese/Chromium/Silicon/Aluminium |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0014 %/Selenium 0.0016 %/Niobium 0.002 %/Cobalt 0.0026 %/Arsenic 0.0098 %/Phosphorus 0.028 %/Silver 0.0292 %/Bismuth 0.035 %/Sulfur 0.041 %/Iron 0.049 %/Antimony 0.103 %/Nickel 0.423 %/Lead 4.9 %/Tin 5.02 %/Zinc 5.09 %/Copper 84.5 %/Oxygen (<0.001) %/Manganese (0.0002) %/Chromium (0.0007) %/Silicon (0.003) %/Aluminium (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction