missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Leaded Bronze, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Leaded Bronze, 50g
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Tellurium/Silver/Cobalt/Bismuth/Zinc/Arsenic/Antimony/Nickel/Tin/Lead/Copper/Aluminium/Silicon/Iron |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Phosphorus 0.003 %/Tellurium 0.01 %/Silver 0.07 %/Cobalt 0.084 %/Bismuth 0.092 %/Zinc 0.11 %/Arsenic 0.165 %/Antimony 0.6 %/Nickel 0.69 %/Tin 8.25 %/Lead 12.5 %/Copper 77.4 %/Aluminium <0.001 %/Silicon <0.001 %/Iron <0.005 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction