missing translation for 'onlineSavingsMsg'
Learn More

Copper Isotopic Standard: Cu @ 10 μg/mL in 2% HNO3 (Enriched in Cu-65)

Artikelnummer. 30314759
Ansicht ändern
Click to view available options
Quantity:
50 mL
Packungsgröße:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
30314759 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30314759 Lieferant VHG Labs Lieferanten-Nr. VHGLIS65CU50
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Copper-65
CAS 14119-06-3
Analyte or Component Names Copper-65
Shipping Condition Room Temperature
Molecular Formula 65 Cu
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym 65 Cu
Solution Type Single Solution
SMILES [65 Cu]
Element Cu
IUPAC Name copper-65
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 64.93
Quantity 50 mL
Formula Weight 64.9278
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.