missing translation for 'onlineSavingsMsg'
Learn More

Copper iodate, anhydrous, 95%

Product Code. 11443780
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443780 50 g 50g
11463780 2 g 2g
11453780 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443780 Supplier Thermo Scientific Alfa Aesar Supplier No. 089459.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13454-89-2 | CuI2O6 | 413.35 g/mol

It used as an intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-89-2
Molecular Formula CuI2O6
Molecular Weight (g/mol) 413.35
MDL Number MFCD00016055
InChI Key LLVVIWYEOKVOFV-UHFFFAOYSA-L
Synonym copper iodate, copper 2+ diiodate, cupric iodate, copper iodate, anhydrous, cu.2io3
PubChem CID 9801829
IUPAC Name copper;diiodate
SMILES [Cu++].[O-][I](=O)=O.[O-][I](=O)=O

Specifications

Melting Point (decomposition)
Color Green
Physical Form Powder
Quantity 50 g
Assay Percent Range 95%
Solubility Information Soluble in water (1.36g/L).
Formula Weight 413.35
Sensitivity Hygroscopic
Appearance Green powder
Density 5.241^1^5
Chemical Name or Material Copper iodate, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.