missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) tungsten oxide, 99.5% (metals basis)

Product Code. 11379367
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379367 50 g 50g
11369367 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379367 Supplier Thermo Scientific Alfa Aesar Supplier No. 040217.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13587-35-4

Copper(II) tungsten oxide nanopowder is used for supercapacitor applications. Nano-sized CuWO4 thin films have been fabricated by radio-frequency (R.F.) sputtering deposition, and are used as positive electrode with both LiClO4 liquid electrolyte and LiPON solid electrolyte in rechargeable lithium batteries. Thin films of copper tungstate is used for nitric oxide sensing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13587-35-4
MDL Number MFCD00016067
Synonym Copper tungstate

Specifications

Physical Form Powder
Quantity 50 g
Merck Index 14,2657
Solubility Information Insoluble in water.
Formula Weight 311.38
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Copper(II) tungsten oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.