Learn More
Copper(II) tungsten oxide, 99.5% (metals basis)
Description
Copper(II) tungsten oxide nanopowder is used for supercapacitor applications. Nano-sized CuWO4 thin films have been fabricated by radio-frequency (R.F.) sputtering deposition, and are used as positive electrode with both LiClO4 liquid electrolyte and LiPON solid electrolyte in rechargeable lithium batteries. Thin films of copper tungstate is used for nitric oxide sensing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 250 g |
| Merck Index | 14,2657 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 311.38 |
| Percent Purity | 99.5% |
| Odor | Odorless |
| Assay | (metals basis) |
| Chemical Name or Material | Copper(II) tungsten oxide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.