missing translation for 'onlineSavingsMsg'
Learn More

Copper (II) trifluoroacetate hydrate, TRC

Product Code. 30363943
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363943 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363943 Supplier Toronto Research Chemicals Supplier No. TRCC685403100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper (II) trifluoroacetate hydrate
CAS 123333-88-0
Molecular Formula 2 C2 F3 O2 . Cu . H2 O
Quantity 100 mg
InChI Formula InChI=1S/2C2HF3O2.Cu.H2O/c2*3-2(4,5)1(6)7;;/h2*(H,6,7);;1H2/q;;+2;/p-2
SMILES O.[Cu+2].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
IUPAC Name copper;2,2,2-trifluoroacetate;hydrate
Molecular Weight (g/mol) 307.592
Formula Weight 306.91
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.