missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) trifluoroacetate hydrate

Product Code. 11455430
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455430 50 g 50g
11445430 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455430 Supplier Thermo Scientific Alfa Aesar Supplier No. 044624.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123333-88-0 | C4CuF6O4 | 289.58 g/mol

As Heterogeneous Catalysts. It is suitable for use as an electrocatalyst in gas diffusion electrodes. As Electrochemical gas sensors. As organic intermediate, as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-88-0
Molecular Formula C4CuF6O4
Molecular Weight (g/mol) 289.58
MDL Number MFCD00013200
InChI Key JIDMEYQIXXJQCC-UHFFFAOYSA-L
Synonym copper ii trifluoroacetate hydrate, 289.58 anhydrous, 2c2hf3o2.cu.xh2o, 2,2,2-trifluoroacetyl oxy cuprio 2,2,2-trifluoroacetate hydrate
PubChem CID 71311159
SMILES [Cu++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Physical Form Crystalline
Quantity 50 g
IUPAC Name copper(2+) ditrifluoroacetate
Formula Weight 289.57 (Anhydrous)
Chemical Name or Material Copper(II) trifluoroacetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.