missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfide, ultra dry, 99.98% (metals basis)

Product Code. 11387867
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387867 10 g 10g
11377867 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387867 Supplier Thermo Scientific Alfa Aesar Supplier No. 014300.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-40-4 | CuS | 95.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-40-4
Molecular Formula CuS
Molecular Weight (g/mol) 95.61
MDL Number MFCD00016066
InChI Key OMZSGWSJDCOLKM-UHFFFAOYSA-N
Synonym copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue
PubChem CID 14831
SMILES [S--].[Cu++]

Specifications

Melting Point 220°C (decomposition)
Physical Form Powder
Quantity 10 g
Merck Index 14,2965
IUPAC Name sulfanylidenecopper
Formula Weight 95.6
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Refractive Index 1.45
Sensitivity Air and moisture sensitive
Density 3.9 g/mL to 4.6 g/mL
Assay (metals basis)
Chemical Name or Material Copper(II) sulfide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.