missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate Solution, Honeywell™

Product Code. 15658480 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Poly bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15658480 25 mL Poly bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15658480 Supplier Honeywell Research Chemicals Supplier No. C228425ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7758-98-7
Physical Form Liquid
Quantity 25 mL
Molecular Formula CuO4S
MDL Number MFCD00010981
UN Number UN3082
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[Cu+2]
IUPAC Name copper;sulfate
Molecular Weight (g/mol) 159.602
PubChem CID 24462
ChEBI CHEBI:23414
Packaging Poly bottle
Chemical Name or Material Copper(II) sulfate solution, 4 % (withv), prepared from copper (II) sulfate pentahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.