missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate pentahydrate, Puriss., suitable for, meets analytical spec. of BP, FCC, Ph. Eur., USP, Solstice
BIOPHARMA

Product Code. 15691210
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
2.5 kg
6 x 500 g
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
100g
1kg
2.5kg
250g
25kg
500g
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15691210 25 kg 25kg
15651210 100 g 100g
15681210 250 g 250g
15601220 500 g 500g
15661210 1 kg 1kg
15671210 2.5 kg 2.5kg
15631220 6 x 500 g Pack of 6
15611220 6 x 1 kg Pack of 6
15621220 6 x 2.5 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15691210 Supplier Honeywell-Fluka Supplier No. 1284925KG
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., suitable for, Meets analytical specification of BP, FCC, Ph. Eur., USP

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 110°C (decomposition)(literature)
CAS Max % 100.0000%
Physical Form Crystals
Quantity 25 kg
Packaging Paper / Poly Bag
Vapor Pressure 7.3mmHg (25°C)
Density 2.28 g/cm3
Linear Formula CuSO4 · 5H2O
UN Number UN3077
IUPAC Name copper;sulfate;pentahydrate
Formula Weight 249.69
Percent Purity 98.5% to 100.5%
Grade puriss.
Chemical Name or Material Copper(II) sulfate pentahydrate, Puriss., suitable for, Meets analytical specification of BP, FCC, Ph. Eur., USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.