missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate pentahydrate, Puriss., suitable for, meets analytical spec. of BP, FCC, Ph. Eur., USP, Honeywell Fluka™
BIOPHARMA

Product Code. 15661210 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
2.5 kg
6 x 500 g
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
100g
1kg
2.5kg
250g
25kg
500g
Pack of 6
This item is not returnable. View return policy

Product Code. 15661210

Brand: Honeywell-Fluka 128491KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., suitable for, Meets analytical specification of BP, FCC, Ph. Eur., USP

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula CuSO4 · 5H2O
UN Number 3077
IUPAC Name copper;sulfate;pentahydrate
Formula Weight 249.69g/mol
Grade puriss.
Vapor Pressure 7.3mmHg (25°C)
Chemical Name or Material Copper(Ii) Sulfate Pentahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Copper(II) sulfate pentahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.