missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate Pentahydrate, Solstice

Product Code. 16615803
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16615803 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615803 Supplier Fluka Supplier No. 3129325KG
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 110°C
CAS 7758-99-8
Color Blue Green
CAS Max % 1
pH 3.5 to 4.5
Quantity 25 kg
Molecular Formula CuH10O9S
Linear Formula CuSO4 · 5 H2O
Synonym Blue Vitriol; Cupric sulfate pentahydrate; Sulfuric acid, copper(II) salt (1:1) pentahydrate
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
IUPAC Name copper(2+) pentahydrate sulfate
Molecular Weight (g/mol) 249.68
Grade ACS
Density 2.28 g/cm3 (20°C)
Chemical Name or Material Copper(II) sulfate pentahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.