Learn More
Copper(II) sulfate pentahydrate, 99%
Description
Copper(II) sulfate, pentahydrate can be added with potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Copper(II) sulfate, pentahydrate can be added with potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling′s solution and Benedict′s solution to test for reducing sugars. Also, used to test protein in Biuret test.
Solubility
Soluble in water and methanol. Insoluble in alcohol.
Notes
Incompatible with steel, finely powdered metals, hydroxylamine, magnesium, hydrazine, nitromethane, alkalies, phosphates and acetylene gas
Specifications
Specifications
| pH | 4 |
| Quantity | 5000 g |
| UN Number | UN3288 |
| Merck Index | 14,2653 |
| Solubility Information | Soluble in water and methanol. Insoluble in alcohol. |
| Formula Weight | 249.68 (159.60 Anhydrous) |
| Percent Purity | 99% |
| Odor | Odorless |
| Density | 2.27 |
| Chemical Name or Material | Copper (II) sulfate pentahydrate |
Safety and Handling
- Copper(II) sulfate pentahydrate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contain: copper (II) sulphatepentahydrate, cupric sulphate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.