missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate pentahydrate, 98%, extra pure

Product Code. 10277593 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Unit Size:
1kg
25kg
5kg
This item is not returnable. View return policy

Product Code. 10277593

Brand: Thermo Scientific Acros 197730250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-99-8 | CuH10O9S | 249.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper;sulfate;pentahydrate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 110.0°C
Color Blue
Physical Form Crystalline Powder and/or Crystals
Quantity 25 kg
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula CuSO4·5H2O
Fieser 01,164; 02,89; 05,162; 06,141; 14,267
Merck Index 15, 2643
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in methanol,slightly soluble in alcohol and glycerol,insoluble in ethanol
Formula Weight 249.68
Percent Purity 98%
Grade Extra Pure
Packaging Fibre drum
Chemical Name or Material Copper(II) sulfate pentahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.