missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11360969
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11360969

Brand: Thermo Scientific Alfa Aesar 010701.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23254-43-5 | CuO4S | 159.60 (anhydrous basis) g/mol

It is used as a catalyst in certain organic transformations like synthesis of ynamides. It is used in Fehling's solution and Benedict's solution for testing reducing sugars, in the Biuret reagent for testing proteins, and in combination with potassium permanganate for testing blood for anemia. It is used in the preparation of Cheshunt compound for horticulture. It finds use as a coloring ingredient in artworks, as an oxidant in combination with potassium permanganate for oxidizing primary alcohols, and as a mordant in dyeing. It is used to etch copper or zinc plates for intaglio print making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23254-43-5
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.60 (anhydrous basis)
MDL Number MFCD00149682
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate monohydrate, cupric sulfate monohydrate, copper sulfate, monohydrate, copper sulfate hydrate, caswell no. 257, epa pesticide chemical code 024402, copper ii sulfate hydrate, sulfuric acid copper 2+ salt 1:1 , monohydrate, copper sulphate hydrate, cuso4.h2o
PubChem CID 61499
IUPAC Name copper;sulfate;hydrate
SMILES [Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Physical Form Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water and methanol. Insoluble in ethanol and most of the organic solvents.
Formula Weight 159.60 (anhydrous)
Odor Odorless
Appearance Blue crystalline aggregrates
Density 2.28g/cm3
Chemical Name or Material Copper(II) sulfate hydrate, m.p. 30° -2H2O, 110° to4H2O, 250° anhydrous; 2.286
compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Copper(II) sulfate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.