missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Sulfate Hydrate, 98%, Honeywell Fluka™

Product Code. 15601900 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15601900 1 kg Poly Bottle 1kg
15621900 5 kg Poly Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15601900 Supplier Honeywell-Fluka Supplier No. 2092011KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

>98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23254-43-5
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.60
MDL Number MFCD00149682
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate monohydrate, cupric sulfate monohydrate, copper sulfate, monohydrate, copper sulfate hydrate, caswell no. 257, epa pesticide chemical code 024402, copper ii sulfate hydrate, sulfuric acid copper 2+ salt 1:1 , monohydrate, copper sulphate hydrate, cuso4.h2o
PubChem CID 61499
IUPAC Name copper;sulfate;hydrate
SMILES [Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 3603g/cu.cm
Color Blue or White
Quantity 1 kg
Linear Formula CuSO4 · xH2O
UN Number UN3077
Formula Weight 159.61g/mol (anhydrous basis)
Percent Purity 98%
Packaging Poly Bottle
Appearance Crystals/Powder
Vapor Pressure 7.3mmHg (25°C)
Density 200°C
Chemical Name or Material Copper(Ii) Sulfate Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.