Learn More
Copper(II) selenide, 99.5% (metals basis)
Description
In semiconductors, as catalyst in Kjeldahl digestion.Copper(II) selenide is involved in the preparation of nanocrystals, which shows significant photothermal heating with a photothermal transduction efficiency, thereby it is used for photothermal therapy applications. It is used as a semiconductor and ideal solar control coating as well as finds application in solar cell technology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 550°C (decomposition) |
| Color | Black |
| Physical Form | Pieces |
| Quantity | 5 g |
| Assay Percent Range | 99.5% (Metals basis) |
| Solubility Information | Soluble in HCl with H2Se evolution,in H2SO4 with SO2 evolution. Oxidized to CuSeO3 by HNO3Soluble in hydrochloric acid,sulfuric acid. Insoluble in water. |
| Formula Weight | 142.5 |
| Density | 5.99 g/cm3 |
| Chemical Name or Material | Copper(II) selenide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.