missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%

Product Code. 11310388
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310388 1 kg 1kg
11320388 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310388 Supplier Thermo Scientific Alfa Aesar Supplier No. 043760.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16570-28-8 | Cu2O7P2 | 301.03 g/mol

Copper(II) pyrophosphate hydrate is used in printed circuit boards. It catalyzes the dimerization of isobutylene to diisobutylene. It is involved in the electrodeposition of copper-zinc alloys based electrolytes. Further, it is used in animal feed as a supplement. In addition to this, it is used in plating of copper on plastics, aluminum, and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16570-28-8
Molecular Formula Cu2O7P2
Molecular Weight (g/mol) 301.03
MDL Number MFCD00150225
InChI Key PEVJCYPAFCUXEZ-UHFFFAOYSA-J
IUPAC Name dicopper(2+) (phosphonatooxy)phosphonate
SMILES [Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Color Blue
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water.
Formula Weight 301.02 (Anhydrous)
Concentration or Composition (by Analyte or Components) P2O7 43.5 to 48.5%
Chemical Name or Material Copper (II) pyrophosphate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.